3-Chlorbenzamidin Hydrochlorid

Lieferant: CHEMPUR

Synonyme: 3-Chlorbenzoesäureamidin Hydrochlorid, 3-Chlorphenylamidin Hydrochlorid

12-0017.1 12-0017.5
CHMP12-0017.1EA 0 EUR
CHMP12-0017.1 CHMP12-0017.5
3-Chlorbenzamidin Hydrochlorid
3-Chlorbenzamidin Hydrochlorid
Formel: C₇H₈Cl₂N₂
Molecular Weight: 191,06 g/mol
MDL: MFCD00276754
CAS-Nummer: 24095-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR