3-Cyanbenzoesäure

Lieferant: COMBI-BLOCKS
Achtung

OS-7665-5G OS-7665-25G OS-7665-100G
COBBOS-7665-5GEA 26 EUR
COBBOS-7665-5G COBBOS-7665-25G COBBOS-7665-100G
3-Cyanbenzoesäure
3-Cyanbenzoesäure
Formel: C₈H₅NO₂
Molecular Weight: 147.13 g/mol
Schmelzpunkt: 219
Lagertemperatur: Raumtemperatur
MDL: MFCD00002486
CAS-Nummer: 1877-72-1
EINECS: 217-511-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR