3-Furonitril 98
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Cyanfuran
L14019.03
L14019.06
L14019.03EA
124
EUR
L14019.03
L14019.06
3-Furonitril 98
3-Furonitril
Purity: 98%
CAS No.: 30078-65-0
Molecular Formula: C5H3NO
Formula Weight: 93.09
Flash Point: 53°(127°F)
Refractive Index: 1.4790
Melting Point: 28-30°
Boiling Point: 150-151°
Beilstein Reference: 507009
UN No.: 2926
MDL No.: MFCD00067042
Formel:
C₅H₃NO Molecular Weight: 93.08 g/mol Siedepunkt: 150…151 °C Schmelzpunkt: 28…30 °C Flammpunkt: 53 °C (127 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067042 CAS-Nummer: 30078-65-0 UN: 2926 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...