3-Acetylphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Acetylphenyl)metaborsäure, Dihydroxy(3-acetylphenyl)boran, 3-Acetylbenzolboronsäure
BB-2186-25G
BB-2186-1G
BB-2186-5G
COBBBB-2186-25GEA
188
EUR
COBBBB-2186-25G
COBBBB-2186-1G
COBBBB-2186-5G
3-Acetylphenylboronsäure
3-Acetylphenylboronsäure
Formel:
C₈H₉BO₃ Molecular Weight: 163,97 g/mol |
MDL:
MFCD01074678 CAS-Nummer: 204841-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...