5-Ethoxy-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (5-Ethoxy-2-fluorphenyl)metaborsäure, Dihydroxy(5-ethoxy-2-fluorphenyl)boran, 5-Ethoxy-2-fluorbenzolboronsäure

BB-8217-5G BB-8217-1G BB-8217-25G
COBBBB-8217-5GEA 204 EUR
COBBBB-8217-5G COBBBB-8217-1G COBBBB-8217-25G
5-Ethoxy-2-fluorphenylboronsäure
5-Ethoxy-2-fluorphenylboronsäure
Formel: C₈H₁₀BFO₃
Molecular Weight: 183.97 g/mol
MDL: MFCD05664321
CAS-Nummer: 900174-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR