3-Ethoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2677-1G BB-2677-100G BB-2677-5G BB-2677-25G
COBBBB-2677-1GEA 37.5 EUR
COBBBB-2677-1G COBBBB-2677-100G COBBBB-2677-5G COBBBB-2677-25G
3-Ethoxyphenylboronsäure
3-Ethoxyphenylboronsäure
Formel: C₈H₁₁BO₃
Molecular Weight: 165,98 g/mol
MDL: MFCD00274219
CAS-Nummer: 90555-66-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR