3-Fluor-4-methylbenzaldehyd

Lieferant: FLUOROCHEM

Synonyme: 3-Fluor-4-tolualdehyd, 3-Fluor-p-tolylaldehyd, 3-Fluor-p-tolualdehyd

005784-5G 005784-25G
FLUO005784-5GEA 80.6 EUR
FLUO005784-5G FLUO005784-25G
3-Fluor-4-methylbenzaldehyd
3-Fluoro-4-methylbenzaldehyde
Formel: C₈H₇FO
Molecular Weight: 138.14 g/mol
MDL: MFCD01631532
CAS-Nummer: 177756-62-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR