3-Fluor-4-nitrobenzoesäure

Lieferant: COMBI-BLOCKS
Achtung

CA-4022-5G CA-4022-25G CA-4022-100G
COBBCA-4022-5GEA 31.2 EUR
COBBCA-4022-5G COBBCA-4022-25G COBBCA-4022-100G
3-Fluor-4-nitrobenzoesäure
3-Fluor-4-nitrobenzoesäure
Formel: C₇H₄FNO₄
Molecular Weight: 185.11 g/mol
MDL: MFCD01862092
CAS-Nummer: 403-21-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR