Methyl-3-fluor-4-nitrobenzoat

Lieferant: COMBI-BLOCKS

Synonyme: 3-Fluor-4-nitrobenzoesäuremethylester

CA-4268-5G CA-4268-1G
COBBCA-4268-5GEA 64 EUR
COBBCA-4268-5G COBBCA-4268-1G
Methyl-3-fluor-4-nitrobenzoat
Methyl-3-fluor-4-nitrobenzoat
Formel: C₈H₆FNO₄
Molecular Weight: 199,14 g/mol
MDL: MFCD08444027
CAS-Nummer: 185629-31-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR