5-Fluor-o-anisaldehyd

Lieferant: CHEMPUR

Synonyme: 5-Fluor-2-anisaldehyd, 5-Fluor-2-methoxybenzaldehyd

FL13325.POR 013325-5G
CHMPFL13325.POREA 0 EUR
CHMPFL13325.POR FLUO013325-5G
5-Fluor-o-anisaldehyd
5-Fluor-o-anisaldehyd
Formel: C₈H₇FO₂
Molecular Weight: 154,14 g/mol
MDL: MFCD00143458
CAS-Nummer: 19415-51-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR