5-Fluor-2-methylbenzolsulfonylchlorid
Lieferant: Thermo Scientific
Synonyme:
5-Fluor-2-methylbenzolsulfonsäurechlorid, 5-Fluor-2-methylbenzol-1-sulfonylchlorid, 5-Fluor-2-methylbenzolsulfochlorid
TL00317.1
TL00317EA
TL00317.50
MAYBTL00317.1EA
0
EUR
MAYBTL00317.1
MAYBTL00317EA
MAYBTL00317.50
5-Fluor-2-methylbenzolsulfonylchlorid
5-Fluor-2-methylbenzolsulfonylchlorid
Formel:
C₇H₆ClFO₂S Molecular Weight: 208,64 g/mol Siedepunkt: 155…158 °C (50 mmHg) |
MDL:
MFCD00052958 CAS-Nummer: 445-05-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...