5-Fluor-2-nitrobenzaldehyd

Lieferant: COMBI-BLOCKS
Achtung

OS-7621-100G OS-7621-5G OS-7621-25G
COBBOS-7621-100GEA 312 EUR
COBBOS-7621-100G COBBOS-7621-5G COBBOS-7621-25G
5-Fluor-2-nitrobenzaldehyd
5-Fluor-2-nitrobenzaldehyd
Formel: C₇H₄FNO₃
Molecular Weight: 169,11 g/mol
MDL: MFCD00153175
CAS-Nummer: 395-81-3
EINECS: 206-903-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR