Methyl-5-fluor-2-nitrobenzoat

Lieferant: COMBI-BLOCKS

Synonyme: 5-Fluor-2-nitrobenzoesäuremethylester

CA-4663-1G CA-4663-5G CA-4663-25G
COBBCA-4663-1GEA 0 EUR
COBBCA-4663-1G COBBCA-4663-5G COBBCA-4663-25G
Methyl-5-fluor-2-nitrobenzoat
Methyl-5-fluor-2-nitrobenzoat
Formel: C₈H₆FNO₄
MDL: MFCD03425619
CAS-Nummer: 393-85-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR