3-Fluorbenzamidin Hydrochlorid

Lieferant: CHEMPUR

Synonyme: 3-Fluorbenzoesäureamidin Hydrochlorid, 3-Fluorphenylamidin Hydrochlorid

12-0049.1 12-0049.5
CHMP12-0049.1EA 0 EUR
CHMP12-0049.1 CHMP12-0049.5
3-Fluorbenzamidin Hydrochlorid
3-Fluorbenzamidin Hydrochlorid
Formel: C₇H₈ClFN₂
Molecular Weight: 174,61 g/mol
MDL: MFCD04114433
CAS-Nummer: 75207-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR