3-Fluorbenzolsulfonamid

Lieferant: FLUOROCHEM

Synonyme: 3-Fluorbenzolsulfonsäureamid

CFF265.5 CFF265.10 017312-5G 017312-1G
CHMPCFF265.5EA 348 EUR
CHMPCFF265.5 CHMPCFF265.10 FLUO017312-5G FLUO017312-1G
3-Fluorbenzolsulfonamid
3-Fluorbenzolsulfonamid
Formel: C₆H₆FNO₂S
Molecular Weight: 175,18 g/mol
MDL: MFCD00042284
CAS-Nummer: 1524-40-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR