3-Fluorphenylboronsäure

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: (3-Fluorphenyl)metaborsäure, 3-Fluorbenzolboronsäure, Dihydroxy(3-fluorphenyl)boran

BB-2290-25G BB-2290-5G BB-2290-100G
COBBBB-2290-25GEA 63.4 EUR
COBBBB-2290-25G COBBBB-2290-5G COBBBB-2290-100G
3-Fluorphenylboronsäure
3-Fluorphenylboronsäure
Formel: C₆H₆BFO₂
Molecular Weight: 139.92 g/mol
Schmelzpunkt: 216…218 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236042
CAS-Nummer: 768-35-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR