Furan-3-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Furanboronic acid, 3-Furanylboronic acid

BB-2310-5G BB-2310-25G BB-2310-1G
COBBBB-2310-5GEA 119 EUR
COBBBB-2310-5G COBBBB-2310-25G COBBBB-2310-1G
Furan-3-boronsäure
Furan-3-boronsäure
Formel: C₄H₅BO₃
Molecular Weight: 111,89 g/mol
MDL: MFCD01319007
CAS-Nummer: 55552-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR