Furan-3-boronsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Furanboronic acid, 3-Furanylboronic acid

L19834.03 L19834.06
L19834.03EA 75.6 EUR
L19834.03 L19834.06
Furan-3-boronsäure ≥97%
Furan-3-boronsäure

Purity: 97% CAS No.: 55552-70-0 Molecular Formula: C4H5BO3 Formula Weight: 111.89 Melting Point: 139-144° Beilstein Reference: 1635653

Formel: C₄H₅BO₃
Molecular Weight: 111,89 g/mol
Schmelzpunkt: 139…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01319007
CAS-Nummer: 55552-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR