Furan-3-boronsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Furanboronic acid, 3-Furanylboronic acid

359401000 359405000 359400025
ACRO359401000EA 21.4 EUR
ACRO359401000 ACRO359405000 ACRO359400025
Furan-3-boronsäure 97%
Furan-3-boronsäure
Formel: C₄H₅BO₃
Molecular Weight: 111,89 g/mol
Schmelzpunkt: 126…128 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01319007
CAS-Nummer: 55552-70-0

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR