3-Hydroxy-4-nitrobenzoesäuremethylester
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl-3-hydroxy-4-nitrobenzoat
CA-4462-5G
CA-4462-25G
CA-4462-1G
COBBCA-4462-5GEA
60.8
EUR
COBBCA-4462-5G
COBBCA-4462-25G
COBBCA-4462-1G
3-Hydroxy-4-nitrobenzoesäuremethylester
Methyl-3-hydroxy-4-nitrobenzoate
Formel:
C₈H₇NO₅ Molecular Weight: 197,15 g/mol |
MDL:
MFCD00016997 CAS-Nummer: 713-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...