3-Hydroxy-2-nitro-6-picolin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
6-Methyl-2-nitro-3-pyridon, 6-Methyl-2-nitropyridin-3-ol, 3-Hydroxy-6-methyl-2-nitropyridin
M-Clarity™ Qualitätsstufe = MQ100
232742-10G
232742-50G
SIAL232742-10GEA
123
EUR
SIAL232742-10G
SIAL232742-50G
3-Hydroxy-2-nitro-6-picolin, Sigma-Aldrich®
3-Hydroxy-2-nitro-6-picolin
Formel:
C₆H₆N₂O₃ Molecular Weight: 154,13 g/mol Schmelzpunkt: 105…106 °C |
MDL:
MFCD00006259 CAS-Nummer: 15128-90-2 EINECS: 239-192-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...