Methyl-3-hydroxybenzoat
Lieferant: COMBI-BLOCKS
Synonyme:
3-Hydroxybenzoesäuremethylester
OT-0751-5G
OT-0751-25G
OT-0751-100G
COBBOT-0751-5GEA
26.2
EUR
COBBOT-0751-5G
COBBOT-0751-25G
COBBOT-0751-100G
Methyl-3-hydroxybenzoat
Methyl-3-hydroxybenzoat
Formel:
C₈H₈O₃ Molecular Weight: 152,15 g/mol |
MDL:
MFCD00002295 CAS-Nummer: 19438-10-9 EINECS: 243-071-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...