3-Isochinolinol

Lieferant: COMBI-BLOCKS

Synonyme: 3-Hydroxyisoquinoline

HC-6650-5G HC-6650-25G HC-6650-1G
COBBHC-6650-5GEA 0 EUR
COBBHC-6650-5G COBBHC-6650-25G COBBHC-6650-1G
3-Isochinolinol
3-Isochinolinol
Formel: C₉H₇NO
MDL: MFCD00075524
CAS-Nummer: 7651-81-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR