3-Hydroxyphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Hydroxybenzolboronsäure, (3-Hydroxyphenyl)metaborsäure, Dihydroxy(3-hydroxyphenyl)boran
BB-2318-1G
BB-2318-25G
BB-2318-5G
COBBBB-2318-1GEA
21.6
EUR
COBBBB-2318-1G
COBBBB-2318-25G
COBBBB-2318-5G
3-Hydroxyphenylboronsäure
3-Hydroxyphenylboronsäure
Formel:
C₆H₇BO₃ Molecular Weight: 137,93 g/mol |
MDL:
MFCD01074603 CAS-Nummer: 87199-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...