3-Indolcarbaldehyd

Lieferant: COMBI-BLOCKS

Synonyme: Indol-3-carboxaldehyd

IN-0171-5G IN-0171-25G IN-0171-100G
COBBIN-0171-5GEA 25.8 EUR
COBBIN-0171-5G COBBIN-0171-25G COBBIN-0171-100G
3-Indolcarbaldehyd
3-Indolcarbaldehyd
Formel: C₉H₇NO
Molecular Weight: 145,16 g/mol
Schmelzpunkt: 192…195 °C
Dichte: 1,34 g/cm³ (20 °C)
MDL: MFCD00005622
CAS-Nummer: 487-89-8
EINECS: 207-665-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR