3-Iodphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8336-5G BB-8336-1G
COBBBB-8336-5GEA 0 EUR
COBBBB-8336-5G COBBBB-8336-1G
3-Iodphenylboronsäure
3-Iodphenylboronsäure
Formel: C₆H₆BIO₂
MDL: MFCD01319015
CAS-Nummer: 221037-98-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR