3-Sulfanyl-1-hexanol 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Mercaptohexan-1-ol
B21965.06
B21965.14
B21965.06EA
59.2
EUR
B21965.06
B21965.14
3-Sulfanyl-1-hexanol 96%
3-Mercapto-1-hexanol
Purity: 96%
CAS No.: 51755-83-0
Molecular Formula: C6H14OS
Formula Weight: 134.24
Flash Point:
Formel:
C₆H₁₄OS Siedepunkt: 250 °C Dichte: 0,974 Flammpunkt: >100 °C (212 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00792515 CAS-Nummer: 51755-83-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...