3-Formylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Boronobenzaldehyd, Dihydroxy(3-formylphenyl)boran, (3-Formylphenyl)metaborsäure, 3-Formylbenzolboronsäure

BB-2304-100G BB-2304-5G BB-2304-25G
COBBBB-2304-100GEA 204 EUR
COBBBB-2304-100G COBBBB-2304-5G COBBBB-2304-25G
3-Formylphenylboronsäure
3-Formylphenylboronsäure
Formel: C₇H₇BO₃
Molecular Weight: 149.94 g/mol
MDL: MFCD00161356
CAS-Nummer: 87199-16-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR