4-Nitro-m-anissäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Methoxy-4-nitrobenzoic acid

CA-4450-5G CA-4450-25G CA-4450-100G
COBBCA-4450-5GEA 36.2 EUR
COBBCA-4450-5G COBBCA-4450-25G COBBCA-4450-100G
4-Nitro-m-anissäure
4-Nitro-m-anissäure
Formel: C₈H₇NO₅
Molecular Weight: 197.15 g/mol
Siedepunkt: 405
Schmelzpunkt: 233
MDL: MFCD00007353
CAS-Nummer: 5081-36-7
EINECS: 225-793-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR