6-Nitro-m-anissäure
Lieferant: COMBI-BLOCKS
Synonyme:
5-Methoxy-2-nitrobenzoic acid
CA-4267-5G
CA-4267-25G
CA-4267-100G
COBBCA-4267-5GEA
28.5
EUR
COBBCA-4267-5G
COBBCA-4267-25G
COBBCA-4267-100G
6-Nitro-m-anissäure
6-Nitro-m-anissäure
Formel:
C₈H₇NO₅ Molecular Weight: 197,15 g/mol Schmelzpunkt: 138 °C |
MDL:
MFCD00151836 CAS-Nummer: 1882-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...