3-Methoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
(3-Methoxyphenyl)metaborsäure, Dihydroxy(3-methoxyphenyl)boran, 3-Methoxybenzolboronsäure
BB-2325-5G
BB-2325-25G
BB-2325-100G
COBBBB-2325-5GEA
25.4
EUR
COBBBB-2325-5G
COBBBB-2325-25G
COBBBB-2325-100G
3-Methoxyphenylboronsäure
3-Methoxyphenylboronsäure
Formel:
C₇H₉BO₃ Molecular Weight: 151,96 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00161359 CAS-Nummer: 10365-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...