3-Methoxypyridin

Lieferant: COMBI-BLOCKS
Achtung

PY-7163-1G PY-7163-25G PY-7163-5G
COBBPY-7163-1GEA 26 EUR
COBBPY-7163-1G COBBPY-7163-25G COBBPY-7163-5G
3-Methoxypyridin
3-Methoxypyridine
Formel: C₆H₇NO
Molecular Weight: 109,13 g/mol
MDL: MFCD00673022
CAS-Nummer: 7295-76-3
EINECS: 230-730-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR