3-Methyl-5-(trifluormethyl)-1H-pyrazol

Lieferant: BIONET RESEARCH

Synonyme: 3-Methyl-5-(trifluormethyl)-pyrazol

11T-0255.500 11T-0255.1 11T-0255.5 11T-0255.10
BIOT11T-0255.500EA 0 EUR
BIOT11T-0255.500 BIOT11T-0255.1 BIOT11T-0255.5 BIOT11T-0255.10
3-Methyl-5-(trifluormethyl)-1H-pyrazol
3-Methyl-5-(trifluoromethyl)-1H-pyrazole
Formel: C₅H₅F₃N₂
Molecular Weight: 150.104 g/mol
MDL: MFCD00829309
CAS-Nummer: 10010-93-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR