3-Methyl-5-(trifluormethyl)-1H-pyrazol
Lieferant: COMBI-BLOCKS
Synonyme:
3-Methyl-5-(trifluormethyl)-pyrazol
HI-1373-5G
HI-1373-25G
HI-1373-1G
COBBHI-1373-5GEA
76.2
EUR
COBBHI-1373-5G
COBBHI-1373-25G
COBBHI-1373-1G
3-Methyl-5-(trifluormethyl)-1H-pyrazol
3-Methyl-5-(trifluoromethyl)-1H-pyrazole
Formel:
C₅H₅F₃N₂ Molecular Weight: 150,104 g/mol |
MDL:
MFCD00829309 CAS-Nummer: 10010-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...