3-Methyl-5-(trifluormethyl)-1H-pyrazol

Lieferant: FLUOROCHEM

Synonyme: 3-Methyl-5-(trifluormethyl)-pyrazol

010332-1G 010332-5G
FLUO010332-1GEA 39.9 EUR
FLUO010332-1G FLUO010332-5G
3-Methyl-5-(trifluormethyl)-1H-pyrazol
3-Methyl-5-(trifluoromethyl)-1H-pyrazole
Formel: C₅H₅F₃N₂
Molecular Weight: 150.104 g/mol
MDL: MFCD00829309
CAS-Nummer: 10010-93-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR