3-Methylbenzamidin Hydrochlorid

Lieferant: CHEMPUR

Synonyme: 3-Methylbenzoesäureamidin Hydrochlorid, 3-Methylphenylamidin Hydrochlorid

12-0029.1 12-0029.5
CHMP12-0029.1EA 0 EUR
CHMP12-0029.1 CHMP12-0029.5
3-Methylbenzamidin Hydrochlorid
3-Methylbenzamidin Hydrochlorid
Formel: C₈H₁₁ClN₂
Molecular Weight: 170.64 g/mol
MDL: MFCD02180878
CAS-Nummer: 20680-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR