3-Tolylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Methylbenzolboronsäure, Dihydroxy(3-methylphenyl)boran, (3-Methylphenyl)metaborsäure, 3-Methylphenylboronsäure
BB-2336-25G
BB-2336-100G
BB-2336-5G
COBBBB-2336-25GEA
76.1
EUR
COBBBB-2336-25G
COBBBB-2336-100G
COBBBB-2336-5G
3-Tolylboronsäure
3-Tolylboronsäure
Formel:
C₇H₉BO₂ Molecular Weight: 135,96 g/mol Schmelzpunkt: 165 °C |
MDL:
MFCD00040198 CAS-Nummer: 17933-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...