3-Nitrobenzamid

Lieferant: Thermo Scientific

Synonyme: 3-Nitrobenzoesäureamid

BTB09266.1 BTB09266.50
MAYBBTB09266.1EA 0 EUR
MAYBBTB09266.1 MAYBBTB09266.50
3-Nitrobenzamid
3-Nitrobenzamid
Formel: C₇H₆N₂O₃
Molecular Weight: 166,14 g/mol
Siedepunkt: 310 °C (1013 hPa)
Schmelzpunkt: 140 °C
MDL: MFCD00007984
CAS-Nummer: 645-09-0
EINECS: 211-431-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR