3-Nitrobenzamid ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3-Nitrobenzoesäureamid
A15481.09
A15481.18
A15481.30
A15481.09EA
30.9
EUR
A15481.09
A15481.18
A15481.30
3-Nitrobenzamid ≥98%
3-Nitrobenzamid
Purity: 98%
CAS No.: 645-09-0
Molecular Formula: C7H6N2O3
Formula Weight: 166.14
Melting Point: 140-143°
Beilstein Reference: 777185
Formel:
C₇H₆N₂O₃ Molecular Weight: 166,14 g/mol Siedepunkt: 310 °C (1013 hPa) Schmelzpunkt: 140…143 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007984 CAS-Nummer: 645-09-0 EINECS: 211-431-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...