3-Nitrobenzamid 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Nitrobenzoesäureamid
183440250
183441000
ACRO183440250EA
55.5
EUR
ACRO183440250
ACRO183441000
3-Nitrobenzamid 98%
3-Nitrobenzamid
Formel:
C₇H₆N₂O₃ Molecular Weight: 166,14 g/mol Siedepunkt: 310 °C (1013 hPa) Schmelzpunkt: 140…144 °C |
MDL:
MFCD00007984 CAS-Nummer: 645-09-0 EINECS: 211-431-0 |
Spezifikation Testergebnisse
Appearance | Yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 140 to 144 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...