3-Nitrobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 3-Nitrobenzolsulfonsäureamid

OT-0123-5G OT-0123-25G
COBBOT-0123-5GEA 28.5 EUR
COBBOT-0123-5G COBBOT-0123-25G
3-Nitrobenzolsulfonamid
3-Nitrobenzolsulfonamid
Formel: C₆H₆N₂O₄S
Molecular Weight: 202,19 g/mol
Schmelzpunkt: 167 °C
MDL: MFCD00007935
CAS-Nummer: 121-52-8
EINECS: 204-477-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR