3-Nitrophenylboronsäure
Lieferant: COMBI-BLOCKS
BB-2347-5G
BB-2347-25G
BB-2347-100G
COBBBB-2347-5GEA
0
EUR
COBBBB-2347-5G
COBBBB-2347-25G
COBBBB-2347-100G
3-Nitrophenylboronsäure
3-Nitrophenylboronsäure
Formel:
C₆H₆BNO₄ Schmelzpunkt: 290…291 °C |
MDL:
MFCD00007193 CAS-Nummer: 13331-27-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...