3-Pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Pyridinylboronsäure, Pyridin-3-boronsäure

BB-2359-5G BB-2359-1G BB-2359-25G BB-2359-100G
COBBBB-2359-5GEA 26.2 EUR
COBBBB-2359-5G COBBBB-2359-1G COBBBB-2359-25G COBBBB-2359-100G
3-Pyridinboronsäure
3-Pyridineboronic acid
Formel: C₅H₆BNO₂
Molecular Weight: 122.92 g/mol
MDL: MFCD00674177
CAS-Nummer: 1692-25-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR