3-Thiophenboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Thienylboronsäure, Thiophen-3-boronsäure
BB-2553-100G
BB-2553-5G
COBBBB-2553-100GEA
362
EUR
COBBBB-2553-100G
COBBBB-2553-5G
3-Thiophenboronsäure
3-Thiopheneboronic acid
Formel:
C₄H₅BO₂S Molecular Weight: 127.96 g/mol Schmelzpunkt: 162…164 °C |
MDL:
MFCD00151851 CAS-Nummer: 6165-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...