4-(Aminomethyl)benzoesäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
B23519.14
B23519.22
B23519.36
B23519.14EA
51.8
EUR
B23519.14
B23519.22
B23519.36
4-(Aminomethyl)benzoesäure 97%
4-(Aminomethyl)benzoic acid
Purity: 97%
CAS No.: 56-91-7
Molecular Formula: C8H9NO2
Formula Weight: 151.16
Melting Point: ca 290° dec.
Beilstein Reference: 1100606
Formel:
C₈H₉NO₂ Molecular Weight: 151,16 g/mol Schmelzpunkt: ∼ 290 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010203 CAS-Nummer: 56-91-7 EINECS: 200-297-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...