4-(Brommethyl)benzolsulfonylchlorid
Lieferant: Thermo Scientific
Synonyme:
4-(Brommethyl)benzolsulfonsäurechlorid, 4-(Brommethyl)benzol-1-sulfonylchlorid, 4-(Brommethyl)benzolsulfochlorid
KM07335.1
KM07335.10
KM07335.50
MAYBKM07335.1EA
0
EUR
MAYBKM07335.1
MAYBKM07335.10
MAYBKM07335.50
4-(Brommethyl)benzolsulfonylchlorid
4-(Brommethyl)benzolsulfonylchlorid
Formel:
C₇H₆BrClO₂S Molecular Weight: 269,55 g/mol Schmelzpunkt: 74…76 °C |
MDL:
MFCD00156129 CAS-Nummer: 66176-39-4 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...