4-(Brommethyl)benzolsulfonylchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-(Brommethyl)benzolsulfonsäurechlorid, 4-(Brommethyl)benzol-1-sulfonylchlorid, 4-(Brommethyl)benzolsulfochlorid
M-Clarity™ Qualitätsstufe = MQ100
557153-1G
557153-5G
SIAL557153-1GEA
30.1
EUR
SIAL557153-1G
SIAL557153-5G
4-(Brommethyl)benzolsulfonylchlorid, Sigma-Aldrich®
4-(Brommethyl)benzolsulfonylchlorid
Formel:
C₇H₆BrClO₂S Molecular Weight: 269,55 g/mol Schmelzpunkt: 74…76 °C |
MDL:
MFCD00156129 CAS-Nummer: 66176-39-4 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...