4-Acetoxy-3-methoxybenzaldehyd ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Vanillinacetat, 4-Formyl-2-methoxyphenylacetat
A12103.14
A12103.22
A12103.30
A12103.14EA
47.1
EUR
A12103.14
A12103.22
A12103.30
4-Acetoxy-3-methoxybenzaldehyd ≥98%
4-Acetoxy-3-methoxybenzaldehyd
Purity: 98%
CAS No.: 881-68-5
Molecular Formula: C10H10O4
Formula Weight: 194.18
Melting Point: 74-78°
Beilstein Reference: 1963795
Formel:
C₁₀H₁₀O₄ Schmelzpunkt: 74…78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003362 CAS-Nummer: 881-68-5 EINECS: 212-920-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...