4-Acetoxybenzaldehyd

Lieferant: FLUOROCHEM

Synonyme: 4-Formylphenylacetat

002485-25G 002485-100G
FLUO002485-25GEA 72.9 EUR
FLUO002485-25G FLUO002485-100G
4-Acetoxybenzaldehyd
4-Acetoxybenzaldehyd
Formel: C₉H₈O₃
Molecular Weight: 164,16 g/mol
MDL: MFCD00003384
CAS-Nummer: 878-00-2
EINECS: 212-898-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR