4-(Hydroxymethyl)benzoesäure
Lieferant: BACHEM BIOCHEMICA
Synonyme:
4-Carboxybenzylalkohol, HMBA
4009594.0005
4009594.0025
BACE4009594.0005EA
0
EUR
BACE4009594.0005
BACE4009594.0025
4-(Hydroxymethyl)benzoesäure
4-(Hydroxymethyl)benzoic acid
Formel:
C₈H₈O₃ Molecular Weight: 152,15 g/mol |
MDL:
MFCD00017598 CAS-Nummer: 3006-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...